Learn More
DWK LIFE SCIENCES™ WHEATON™ Standard Roller Bottom Drive System Options with Belt Break Alarm and Battery Backup
Shop All DWK Life Sciences Products
Description
- Field installation options package for Wheaton™ Standard Roller Bottom Drive systems
- Includes belt break alarm to alert users to drive belt failure
- Automatic battery backup helps maintain operation during power interruptions
- Designed for compatibility with Wheaton™ Bottom-Drive Standard Roller Apparatus
- Supports modular and production spaced roller system configurations
Specifications
Specifications
| Product Type | Installation Kit |
| For Use With (Equipment) | Wheaton Bottom-drive Standard Roller Apparatus |
| Includes | Belt break alarm and automatic battery backup |