Learn More
DWK LIFE SCIENCES™ WHEATON™ Field Installation Options Package for WHEATON™ Standard Roller Top Drive System
SDP
Catalog No. 02924964 Shop All DWK Life Sciences Products
Change view
Click to view available options
For Use With (Equipment):
Wheaton Top-drive Standard Roller Apparatus
Catalog No. For Use With (Equipment)
02-924-964 Wheaton Top-drive Standard Roller Apparatus
1 options
Catalog No. 02-924-964 Supplier DWK LIFE SCIENCES™ Supplier No. W348890V2
Add to Cart
Add to Cart
WHEATON™ Field Installation Options Package for WHEATON™ Standard Roller Top Drive System adds belt break alarm and automatic battery backup to your existing Standard Roller Top Drive apparatus. Upgrade your roller system with essential protection and power continuity features.
  • Field installation options package for the Wheaton Standard Roller Top Drive System
  • Includes belt break alarm to alert users to drive belt failure
  • Automatic battery backup ensures continued operation during power interruptions
  • Designed for use with the Wheaton Standard Roller Top Drive roller apparatus
  • Supplied by DWK Life Sciences under the Wheaton brand
TRUSTED_SUSTAINABILITY
Specifications
Product Type Installation Kit
For Use With (Equipment) Wheaton Top-drive Standard Roller Apparatus
Includes Belt break alarm and automatic battery backup