Learn More
Electron Microscopy Sciences Instant Readout Digital Caliper
SDP
Supplier:  Electron Microscopy Sciences 6206540
Encompass_Preferred
This caliper is made from stainless steel and measures inside and outside in inches and millimeters. Read-out from 0 to 6 inches with 0.001" resolution, or 0 to 150 mm with 0.01 mm resolution. Includes a zero set button, depth measuring rod and a fine adjustment thumb wheel. A protective carrying case and battery are included.
Catalog No. 50-996-491