Learn More
MSC SC Johnson Professional Foaming Hand Sanitizer, Non-Alcohol, 1.2 L, 3/case
Catalog No. 502868687
Encompass
This non-alcohol based foaming hand sanitizer is highly effective, perfume-free, and dye-free, designed for critical hand hygiene in environments where alcohol is a concern. The foam formula provides user control, allows for more frequent hand hygiene moments, and includes a skin conditioner to prevent dryness.
  • Non-alcohol based for risk-sensitive areas
  • Contains benzalkonium chloride for antimicrobial efficacy up to 99.999%
  • Enriched with aloe vera to prevent drying and leave skin smooth
  • Perfume-free and dye-free for sensitive users
  • Does not contain gelling agents to avoid sticky residue
  • Pleasant foam for frequent use, encouraging compliance
  • Mild preservative reduces risk of skin irritation
  • NSF E-3 rated, complying with NSF International standards
Catalog No. 50-286-8687 Supplier MSC Supplier No. 44343499
This non-alcohol based foaming hand sanitizer is highly effective, perfume-free, and dye-free, designed for critical hand hygiene in environments where alcohol is a concern. The foam formula provides user control, allows for more frequent hand hygiene moments, and includes a skin conditioner to prevent dryness.
  • Non-alcohol based for risk-sensitive areas
  • Contains benzalkonium chloride for antimicrobial efficacy up to 99.999%
  • Enriched with aloe vera to prevent drying and leave skin smooth
  • Perfume-free and dye-free for sensitive users
  • Does not contain gelling agents to avoid sticky residue
  • Pleasant foam for frequent use, encouraging compliance
  • Mild preservative reduces risk of skin irritation
  • NSF E-3 rated, complying with NSF International standards