Learn More
Agilent Technologies INSTANTPC CLEANUP MODULE,96-CT
Catalog No. NC2292354
Encompass
AdvanceBio Gly-X InstantPC cleanup module, 96-ct (formerly ProZyme). Module for use with Gly-X glycan prep and InstantPC labeling modules.
This item is not returnable. View return policy
Catalog No. NC2292354 Supplier Agilent Technologies Supplier No. GX96102
AdvanceBio Gly-X InstantPC cleanup module, 96-ct (formerly ProZyme). Module for use with Gly-X glycan prep and InstantPC labeling modules.