Learn More
Ace Glass, Inc. Instatherm; oil bath, 1200mL, 142mm ID, 150mm OD, 75mm height, 120v/5amp max, 12ft PTFE-clad, 2-prong, permanently attached controller power cord.
SDP
Catalog No. 5097770
Encompass
Instatherm; oil bath, 1200mL, 142mm ID, 150mm OD, 75mm height, 120v/5amp max, 12ft PTFE-clad, 2-prong, permanently attached controller power cord.
Catalog No. 50-977-70 Supplier Ace Glass, Inc. Supplier No. 960215
Instatherm; oil bath, 1200mL, 142mm ID, 150mm OD, 75mm height, 120v/5amp max, 12ft PTFE-clad, 2-prong, permanently attached controller power cord.