Learn More
Ace Glass, Inc. Instatherm; oil bath, 160mL, 64mm ID, 70mm OD, 50mm height, 20v/5amp max, 12ft PTFE-clad, 2-prong, permanently attached controller power cord.
SDP
Catalog No. 5097768
Encompass
Instatherm; oil bath, 160mL, 64mm ID, 70mm OD, 50mm height, 20v/5amp max, 12ft PTFE-clad, 2-prong, permanently attached controller power cord.
Catalog No. 50-977-68 Supplier Ace Glass, Inc. Supplier No. 960207
Instatherm; oil bath, 160mL, 64mm ID, 70mm OD, 50mm height, 20v/5amp max, 12ft PTFE-clad, 2-prong, permanently attached controller power cord.