Learn More
Ace Glass, Inc. Instatherm; oil bath, low form, 700mL, 119mm ID, 125mm OD, 65mm height, includes 9698-20 connecting controller cord and9601-30 thermocouple/thermometer clip, 40v/8amp max
SDP
Catalog No. 5097764
Encompass
Instatherm; oil bath, low form, 700mL, 119mm ID, 125mm OD, 65mm height, includes 9698-20 connecting controller cord and9601-30 thermocouple/thermometer clip, 40v/8amp max
Catalog No. 50-977-64 Supplier Ace Glass, Inc. Supplier No. 960144
Instatherm; oil bath, low form, 700mL, 119mm ID, 125mm OD, 65mm height, includes 9698-20 connecting controller cord and9601-30 thermocouple/thermometer clip, 40v/8amp max