Learn More
Ace Glass, Inc. Permanent cord Instatherm; oil bath kit consists of (1)each 340mL, 700mL & 1200mL baths, thermocouple/thermometer holding clips for each bath, (1) 102mm inconel J type probe and (1) 12324-25 temperature controller
SDP
Catalog No. 5098098
Encompass
Permanent cord Instatherm; oil bath kit consists of (1)each 340mL, 700mL & 1200mL baths, thermocouple/thermometer holding clips for each bath, (1) 102mm inconel J type probe and (1) 12324-25 temperature controller
Catalog No. 50-980-98 Supplier Ace Glass, Inc. Supplier No. 9602364
Permanent cord Instatherm; oil bath kit consists of (1)each 340mL, 700mL & 1200mL baths, thermocouple/thermometer holding clips for each bath, (1) 102mm inconel J type probe and (1) 12324-25 temperature controller