Learn More
Ace Glass, Inc. Instatherm; griffin beaker with lip and pouring spout, 400mL, 72mm ID, 77mm OD, 110mm height, 40v/6amp max, includes9698-20 connecting controller cord
SDP
Catalog No. 5097780
Encompass
Instatherm; griffin beaker with lip and pouring spout, 400mL, 72mm ID, 77mm OD, 110mm height, 40v/6amp max, includes9698-20 connecting controller cord
Catalog No. 50-977-80 Supplier Ace Glass, Inc. Supplier No. 960542
Instatherm; griffin beaker with lip and pouring spout, 400mL, 72mm ID, 77mm OD, 110mm height, 40v/6amp max, includes9698-20 connecting controller cord