Learn More
Ace Glass, Inc. Flask, Instatherm, round bottom, 3000mL, two necks, 29/42 joints, 120v/10amp max
SDP
Catalog No. 5097791
Encompass
Flask, Instatherm, round bottom, 3000mL, two necks, 29/42 joints, 120v/10amp max
Catalog No. 50-977-91 Supplier Ace Glass, Inc. Supplier No. 9637170
Flask, Instatherm, round bottom, 3000mL, two necks, 29/42 joints, 120v/10amp max