Learn More
Ace Glass, Inc. Instatherm; oil bath, low form, 1.2 liter, 142mm ID, 150mm OD, 75mm height, includes 9698-20 connecting controller cord, 120v/5amp max
SDP
Catalog No. 50977057
Encompass
Instatherm; oil bath, low form, 1.2 liter, 142mm ID, 150mm OD, 75mm height, includes 9698-20 connecting controller cord, 120v/5amp max
Catalog No. 50-977-057 Supplier Ace Glass, Inc. Supplier No. 960116
Instatherm; oil bath, low form, 1.2 liter, 142mm ID, 150mm OD, 75mm height, includes 9698-20 connecting controller cord, 120v/5amp max