Learn More
Ace Glass, Inc. Instatherm; oil bath, low form, 2.6 liter, 182mm ID, 190mm OD, 100mm height, includes 9698-20 connecting controller cord, 120v/10amp max
SDP
Catalog No. 50980092
Encompass
Instatherm; oil bath, low form, 2.6 liter, 182mm ID, 190mm OD, 100mm height, includes 9698-20 connecting controller cord, 120v/10amp max
Catalog No. 50-980-092 Supplier Ace Glass, Inc. Supplier No. 960118
Instatherm; oil bath, low form, 2.6 liter, 182mm ID, 190mm OD, 100mm height, includes 9698-20 connecting controller cord, 120v/10amp max