Learn More
Ace Glass, Inc. Instatherm; oil bath, immerses 300mL flasks and smaller, 90mm ID, 100mm OD, 100mm height, 40v/10amp max, includes 9698-20connecting controller cord
SDP
Catalog No. 5097776
Encompass
Instatherm; oil bath, immerses 300mL flasks and smaller, 90mm ID, 100mm OD, 100mm height, 40v/10amp max, includes 9698-20connecting controller cord
Catalog No. 50-977-76 Supplier Ace Glass, Inc. Supplier No. 960302
Instatherm; oil bath, immerses 300mL flasks and smaller, 90mm ID, 100mm OD, 100mm height, 40v/10amp max, includes 9698-20connecting controller cord