Learn More
Ace Glass, Inc. Instatherm; oil bath, immerses 3L flasks and smaller, 200mm ID, 210mm OD, 200mm height, 120v/10amp max, includes 9698-20connecting controller cord and thermocouple/thermometer clip
SDP
Catalog No. NC1418475
Encompass
Instatherm; oil bath, immerses 3L flasks and smaller, 200mm ID, 210mm OD, 200mm height, 120v/10amp max, includes 9698-20connecting controller cord and thermocouple/thermometer clip
Catalog No. NC1418475 Supplier Ace Glass, Inc. Supplier No. 960324
Instatherm; oil bath, immerses 3L flasks and smaller, 200mm ID, 210mm OD, 200mm height, 120v/10amp max, includes 9698-20connecting controller cord and thermocouple/thermometer clip