Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Seward™ Insterprep™ Himedia™ Buffered Peptone Water (Bag, Granules 225mL)

Catalog No. 13874762
Change view
Click to view available options
Quantity:
225 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
13-874-762 225 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 13-874-762 Supplier Seward™ Supplier No. INS2100

Designed to speed up and simplify sample preparation for microbiological analysis

  • Instantly soluble, sterile
  • Granulated media
  • Ready for instant use
  • Reduces costs by cutting preparation time and reallocating resources
  • Minimize waste by just preparing what you need
  • Eliminate bottlenecks
  • Reliable sample preparation
  • Visible traceability
  • Used as pre-enrichment medium for increasing recovery of injured Salmonella species from foods prior to selective enrichment and isolation

Certifications

The composition and performance criteria of this medium are as per the applications laid down in ISO 6579-2002.

Specifications

Content And Storage Store below 30°C in tightly closed container and the prepared medium between 2° to 8°C.
Form Liquid
Product Type Buffered Peptone Water
Format Bag
For Use With (Application) Pre-enrichment medium
pH 6.8 to 7.2
Quantity 225 mL
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.