Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Seward™ Insterprep™ Himedia™ Fraser Broth w/Supplement, Granulated Bag

Catalog No. 13874764
Change view
Click to view available options
Quantity:
225 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
13-874-764 225 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 13-874-764 Supplier Seward™ Supplier No. INS2102

For selective enrichment of Listeria species from foods

  • Instantly soluble, sterile
  • Granulated media
  • Ready for instant use
  • Reduces costs by cutting preparation time and reallocating resources
  • Minimize waste by just preparing what you need
  • Eliminate bottlenecks
  • Reliable sample preparation
  • Visible traceability
  • Appearance: Cream to yellow homogeneous granular powder
  • Colour and Clarity of prepared medium: Fluorescent yellow coloured clear solution

Certifications

The composition and performance criteria of this medium are as per the applications laid down in ISO 6579-2002

Specifications

Content And Storage Store dehydrated powder in tightly closed container and prepared medium at 2° to 8°C. Use before expiry date on the label
Product Type Fraser Broth
Format Bag
For Use With (Application) Selective enrichment of Listeria species from foods
pH 7 to 7.4
Quantity 225 mL
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.