Learn More
Advanced Instruments Inc InstiGRO CHO PLUS, Enhances Clonal Growth, Animal Component-Free, 25 mL
SDP
Catalog No. NC2313444
Encompass
InstiGRO CHO PLUS 25 mL is an animal component-free cell culture supplement designed to enhance the growth and cloning efficiency of suspension CHO cells. Its advanced formulation supports single-cell isolation and promotes robust clonal colonies while maintaining optical clarity for imaging systems. Ideal for improving cloning efficiency, it streamlines cell line development projects, potentially reducing plate usage and enabling earlier clonal expansion. This product ensures rigorous quality control and is suitable for research use only.
Catalog No. NC2313444 Supplier Advanced Instruments Inc Supplier No. RS1225
InstiGRO CHO PLUS 25 mL is an animal component-free cell culture supplement designed to enhance the growth and cloning efficiency of suspension CHO cells. Its advanced formulation supports single-cell isolation and promotes robust clonal colonies while maintaining optical clarity for imaging systems. Ideal for improving cloning efficiency, it streamlines cell line development projects, potentially reducing plate usage and enabling earlier clonal expansion. This product ensures rigorous quality control and is suitable for research use only.