Learn More
Advanced Instruments Inc InstiGRO HEK Cell Culture Supplement, Enhances Clonal Outgrowth, 20x Concentrated, 25 mL
SDP
Catalog No. NC3356452
Encompass
InstiGRO HEK - 25 mL is a premium, chemically defined cell culture supplement designed for enhanced growth and cloning efficiency of suspension HEK cells. This 20x concentrated solution supports cell viability and robust outgrowth during isolation, offering compatibility with imaging-based workflows. It's free from animal components, easy to use, and manufactured under ISO 9001 standards, making it ideal for research applications.
Catalog No. NC3356452 Supplier Advanced Instruments Inc Supplier No. RS1325
InstiGRO HEK - 25 mL is a premium, chemically defined cell culture supplement designed for enhanced growth and cloning efficiency of suspension HEK cells. This 20x concentrated solution supports cell viability and robust outgrowth during isolation, offering compatibility with imaging-based workflows. It's free from animal components, easy to use, and manufactured under ISO 9001 standards, making it ideal for research applications.