Learn More
Advanced Instruments Inc InstiSHAKE™ HEK, Enhances Cell Viability, 20x Concentrated Solution, 25 mL
SDP
Supplier:  Advanced Instruments Inc RS2325
Encompass
InstiSHAKE HEK is a 20x concentrated solution that enhances the viability and density of HEK293 cell lines, specifically aiding their transition from static to shaking culture. This chemically defined, animal component-free supplement nurtures the health of bulk cultures, making it ideal for scalable cell culture manufacturing. With an 18-month shelf life and storage at 2-8°C, its ease of use simplifies integration into cell culture workflows.
Catalog No. NC3356453