Learn More
Med Vet International Miltex Dental Excavator, #38/39, Double-Ended, Octagonal Handle
Catalog No. 502830593
Encompass_Preferred
The Miltex 70-10 38/39 dental excavator is made of OR grade German stainless steel and is double-ended with an octagonal handle. Excavators are used to remove caries dentin and other dental materials (temporary cement, etc.).
Catalog No. 50-283-0593 Supplier Med Vet International Supplier No. MIL7010
The Miltex 70-10 38/39 dental excavator is made of OR grade German stainless steel and is double-ended with an octagonal handle. Excavators are used to remove caries dentin and other dental materials (temporary cement, etc.).