Learn More
Med Vet International Instrument Lubricant Concentrate 1 gallon, Concentrated
Catalog No. 502522369
Encompass_Preferred
Surgical Instrument Lubricant Milk is used after every use of the instrument. Not only does this product lubricate all moving parts, but it helps prevent spotting, staining and rusting of your most precious instruments. This product is not a detergent or sterilizer. Instruments must be autoclaved after Surgical Instrument Lubricant Milk has been applied. The lubricant bath should be replaced at least weekly or more frequently depending on number of instruments processed. Ingredients: Water Polybutene Oleth-2 Oleic Acid Triethanolamine Methylparaben Propylparaben Hydrogenated Castol Oil One gallon makes SEVEN GALLONS of lubricant
Catalog No. 50-252-2369 Supplier Med Vet International Supplier No. ILG
 Restricted Item
Add to Cart
Add to Cart
Surgical Instrument Lubricant Milk is used after every use of the instrument. Not only does this product lubricate all moving parts, but it helps prevent spotting, staining and rusting of your most precious instruments. This product is not a detergent or sterilizer. Instruments must be autoclaved after Surgical Instrument Lubricant Milk has been applied. The lubricant bath should be replaced at least weekly or more frequently depending on number of instruments processed. Ingredients: Water Polybutene Oleth-2 Oleic Acid Triethanolamine Methylparaben Propylparaben Hydrogenated Castol Oil One gallon makes SEVEN GALLONS of lubricant