Learn More
Inorganic Ventures INSTR PERFORM CHECK 2 125mL
SDP
Catalog No. 50723862
Encompass_Preferred
Matrix: Nitric Acid / Hydrofluoric Acid (HNO3 / HF), Dilution: 1:100, Analyte: Sb, Ti, Mo, and Sn 200 ug/mL each, and SiO2 1000 ug/mL.
ATTENTION: Inorganic Ventures is no longer supplying paper copies of Certificate of Analyses(CoA) and Safety Data Sheets (SDS). Please use this link to download your copies: https://www.inorganicventures.com/coa-sds-search
Catalog No. 50-723-862 Supplier Inorganic Ventures Supplier No. WWIPC2125ML
Matrix: Nitric Acid / Hydrofluoric Acid (HNO3 / HF), Dilution: 1:100, Analyte: Sb, Ti, Mo, and Sn 200 ug/mL each, and SiO2 1000 ug/mL.
ATTENTION: Inorganic Ventures is no longer supplying paper copies of Certificate of Analyses(CoA) and Safety Data Sheets (SDS). Please use this link to download your copies: https://www.inorganicventures.com/coa-sds-search