Learn More
Med Vet International Instrument Tip Guard Comply 9-1/2 L X 5-1/2 W inch, Clear, Plastic, Rigid Paperboard With Pouch, 13915, 100/Box
Catalog No. 502829673
Encompass_Preferred
Dual chemical indicator bars are printed on paper for steam and eo sterilization monitoring The chemical indicator is used for both exposure control monitoring and pack control monitoring
Catalog No. 50-282-9673 Supplier Med Vet International Supplier No. 155495BX
Dual chemical indicator bars are printed on paper for steam and eo sterilization monitoring The chemical indicator is used for both exposure control monitoring and pack control monitoring