Learn More
Roboz Surgical INSTRMT TRAY LID 10 X 6.5
SDP
Catalog No. NC9453140
Encompass
Instrument Tray Lid; 10 X 6.5 in. These Trays Are Electro-Polished To Provide A Tough, Corrosion-Resistant Surface And A Bright, Clean Look.Constructed Of Heavy-Duty, 304 18-8 Stainless Steel, Pmp Trays Are Available In Many Different Styles And Sizes T
Catalog No. NC9453140 Supplier Roboz Surgical Supplier No. RT1401L
Instrument Tray Lid; 10 X 6.5 in. These Trays Are Electro-Polished To Provide A Tough, Corrosion-Resistant Surface And A Bright, Clean Look.Constructed Of Heavy-Duty, 304 18-8 Stainless Steel, Pmp Trays Are Available In Many Different Styles And Sizes T