Learn More
Grainger Instructional Label Care PK250
Catalog No. 502764996
Encompass_Preferred
The Roll Products Chemical Label is a 4" x 6" label made of white vinyl with a permanent adhesive backing. It is designed for identifying chemical containers and includes space to fill in the chemical name, signal word, hazard statement, and pictogram. The vinyl material is chemical and water resistant. Each pack contains 100 labels. This is a useful product for workplaces that need OSHA-compliant chemical labeling.
Catalog No. 50-276-4996 Supplier Grainger Supplier No. 16U838
The Roll Products Chemical Label is a 4" x 6" label made of white vinyl with a permanent adhesive backing. It is designed for identifying chemical containers and includes space to fill in the chemical name, signal word, hazard statement, and pictogram. The vinyl material is chemical and water resistant. Each pack contains 100 labels. This is a useful product for workplaces that need OSHA-compliant chemical labeling.