Learn More
Grainger Instructional Label Care PK250
Catalog No. 502765240
Encompass_Preferred
The Roll Products Chemical Label is a 2" x 4" adhesive label made of white polyester with a permanent acrylic adhesive. It is designed for identifying chemical containers and resist chemicals, moisture, and abrasion. The labels can withstand temperatures from -40°F to 250°F. They come in packs of 500 labels.
Catalog No. 50-276-5240 Supplier Grainger Supplier No. 16U836
The Roll Products Chemical Label is a 2" x 4" adhesive label made of white polyester with a permanent acrylic adhesive. It is designed for identifying chemical containers and resist chemicals, moisture, and abrasion. The labels can withstand temperatures from -40°F to 250°F. They come in packs of 500 labels.