Learn More
Grainger Instructional Label Care PK500
Catalog No. 502765849
Encompass_Preferred
The Grainger Shipping Labels are 2" x 4" thermal labels for use with thermal printers. Made of white paper with a permanent acrylic adhesive, they come in packs of 500. These blank labels allow users to print shipping and address information and adhere to various materials like cardboard, kraft paper, coated paper, glass, and plastics. Ideal for small to medium packages, the thermal printing technology ensures fast, high-quality printing without ink. These versatile labels are perfect for warehouse, office, or industrial shipping needs.
Catalog No. 50-276-5849 Supplier Grainger Supplier No. 16U833
The Grainger Shipping Labels are 2" x 4" thermal labels for use with thermal printers. Made of white paper with a permanent acrylic adhesive, they come in packs of 500. These blank labels allow users to print shipping and address information and adhere to various materials like cardboard, kraft paper, coated paper, glass, and plastics. Ideal for small to medium packages, the thermal printing technology ensures fast, high-quality printing without ink. These versatile labels are perfect for warehouse, office, or industrial shipping needs.