Learn More
Grainger Instructional Label Care PK500
Catalog No. 502768446
Encompass_Preferred
The Grainger Shipping Labels are 2" x 4" thermal transfer labels designed for shipping and warehouse applications. They are made of white paper with a permanent acrylic adhesive and are compatible with thermal transfer printers. Each roll contains 2000 labels with no liner and measures 2" diameter. The operating temperature range is -40°F to 160°F. These shipping labels meet UPS and FedEx requirements and are ideal for shipping, warehouse labeling, and inventory management.
Catalog No. 50-276-8446 Supplier Grainger Supplier No. 16U873
The Grainger Shipping Labels are 2" x 4" thermal transfer labels designed for shipping and warehouse applications. They are made of white paper with a permanent acrylic adhesive and are compatible with thermal transfer printers. Each roll contains 2000 labels with no liner and measures 2" diameter. The operating temperature range is -40°F to 160°F. These shipping labels meet UPS and FedEx requirements and are ideal for shipping, warehouse labeling, and inventory management.