Learn More
Grainger Instructional Label Care PK500
Catalog No. 502765088
Encompass_Preferred
The Grainger 3CLY1 Shipping Labels are 2" x 4" thermal transfer labels designed for shipping and warehouse applications. They are made from a white polyester material with a permanent acrylic adhesive and are compatible with thermal transfer printers. Each roll contains 1,500 labels and 12 rolls per case. The labels have a service temperature range of -65°F to 200°F. They meet UPS and FedEx requirements for shipping labels. The labels resist moisture, abrasion, and chemicals. They provide a cost-effective solution for shipping, warehouse labeling, distribution centers, and fulfillment operations.
Catalog No. 50-276-5088 Supplier Grainger Supplier No. 16U850
The Grainger 3CLY1 Shipping Labels are 2" x 4" thermal transfer labels designed for shipping and warehouse applications. They are made from a white polyester material with a permanent acrylic adhesive and are compatible with thermal transfer printers. Each roll contains 1,500 labels and 12 rolls per case. The labels have a service temperature range of -65°F to 200°F. They meet UPS and FedEx requirements for shipping labels. The labels resist moisture, abrasion, and chemicals. They provide a cost-effective solution for shipping, warehouse labeling, distribution centers, and fulfillment operations.