Learn More
Grainger Instructional Label Care PK500
Catalog No. 502765664
Encompass_Preferred
The Grainger 3CLY1 Shipping Labels are 2" x 4" thermal transfer labels designed for shipping and warehouse applications. They are made from a white polyester material with a permanent acrylic adhesive and are compatible with thermal transfer printers. Each roll contains 1,000 labels and measures 2.25" diameter with a 1" core. The operating temperature range is -40°F to 160°F. These shipping labels meet UPS and FedEx requirements and are ideal for shipping, warehouse labeling, and inventory management.
Catalog No. 50-276-5664 Supplier Grainger Supplier No. 16U896
The Grainger 3CLY1 Shipping Labels are 2" x 4" thermal transfer labels designed for shipping and warehouse applications. They are made from a white polyester material with a permanent acrylic adhesive and are compatible with thermal transfer printers. Each roll contains 1,000 labels and measures 2.25" diameter with a 1" core. The operating temperature range is -40°F to 160°F. These shipping labels meet UPS and FedEx requirements and are ideal for shipping, warehouse labeling, and inventory management.