Learn More
Grainger Instructional Label Contents PK500
Catalog No. 502767304
Encompass_Preferred
The Grainger 16U889 Instructional Label Contents is a pack of 500 instructional labels. These labels are 2" x 3" in size and are made from a vinyl material with a permanent acrylic adhesive backing. The labels have a white background with black text that reads "Contents" and have space below to write in the contents. They can adhere to a variety of surfaces including plastic, glass, and metal. With 500 labels per pack, this provides a versatile labeling solution for inventory organization and identification in warehouses, factories, distribution centers, and more.
Catalog No. 50-276-7304 Supplier Grainger Supplier No. 16U889
The Grainger 16U889 Instructional Label Contents is a pack of 500 instructional labels. These labels are 2" x 3" in size and are made from a vinyl material with a permanent acrylic adhesive backing. The labels have a white background with black text that reads "Contents" and have space below to write in the contents. They can adhere to a variety of surfaces including plastic, glass, and metal. With 500 labels per pack, this provides a versatile labeling solution for inventory organization and identification in warehouses, factories, distribution centers, and more.