Learn More
Grainger Instructional Label Contents PK500
Supplier: Grainger 16U874
The Tapecase Shipping Labels are 3" high x 2.625" wide pressure sensitive labels on a roll with 500 labels per roll. The labels have a red header and white body. They are made from thermal transfer materials and are suitable for use in thermal transfer printers. The adhesive is permanent acrylic that adheres to corrugated cardboard and other shipping materials.