Learn More
Perkin Elmer US LLC Instrument Calibration Standard 2, 5% HNO3
Catalog No. 509045836
Encompass_Preferred
Concentration of Element includes: Concentration Content 100 µg/mL Ag, Al, As, Ba, Be, Ca, Cd, Co, Cr, Cu, Fe, K, Mg, Mn, Mo, Na, Ni, Pb, Sb, Se, Sn, Sr, Ti, Tl, V, Zn
Catalog No. 50-904-5836 Supplier Perkin Elmer US LLC Supplier No. N9301721
Concentration of Element includes: Concentration Content 100 µg/mL Ag, Al, As, Ba, Be, Ca, Cd, Co, Cr, Cu, Fe, K, Mg, Mn, Mo, Na, Ni, Pb, Sb, Se, Sn, Sr, Ti, Tl, V, Zn