Learn More
Sklar Instruments Instrument Cassette with Spring Holding, Sklar, 220mm x 125mm x 40mm, Removable Locking Lid, Perforated Sides. EA/1.
SDP
Catalog No. 501437294 Shop All Sklar Instruments Products
Encompass_Preferred
Instrument Cassette with Spring Holding, Sklar, 220mm x 125mm x 40mm, Removable Locking Lid, Perforated Sides. Sklar Instrument Cassettes are used for cleaning, storing, and sterilizing instruments. The silicone supports are repositionable to assist in arranging and stabilizing instruments. Cassettes are available with a hinged or detachable lid. The baskets have clamps to secure to bottom of cassette when the device is being processed.The over all size is 8-3/4 x 5 x 1-1/2 in.
This item is not returnable. View return policy
Catalog No. 50-143-7294 Supplier Sklar Instruments Supplier No. 106740
Instrument Cassette with Spring Holding, Sklar, 220mm x 125mm x 40mm, Removable Locking Lid, Perforated Sides. Sklar Instrument Cassettes are used for cleaning, storing, and sterilizing instruments. The silicone supports are repositionable to assist in arranging and stabilizing instruments. Cassettes are available with a hinged or detachable lid. The baskets have clamps to secure to bottom of cassette when the device is being processed.The over all size is 8-3/4 x 5 x 1-1/2 in.