Learn More
Stoelting™ Instrument Cup
SDP
Catalog No. 10000861 Shop All Stoelting Co Products
Change view
Click to view available options
Size:
Small
Large
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Size
10-000-861 Small
10-000-859 Large
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 10-000-861 Supplier Stoelting™ Supplier No. 52203
Add to Cart
Add to Cart
Stoelting™ Instrument Cup replaces glass and metal instrument pans, and won't damage delicate instrument tips.
Large instruments rest against the bottom and side, while small instruments may be suspended from the scalloped brim.
Specifications
For Use With (Application) Neuroscience
Size Small