Learn More
World Precision Instrument Instrument Demagnetizer
SDP
Catalog No. 502521380
Encompass
Instantly magnetize or demagnetize small surgical instruments and small steel parts that are over 1.3" in length and 0.125" in diameter. No batteries or electricity required. The box is high quality molded ABS plastic, impact resistant and cadmium free. Dimensions are 2" x 2" x1" The innovative step design lets you control the level of magnetism imposed on your tools.
Catalog No. 50-252-1380 Supplier World Precision Instrument Supplier No. 501900
Instantly magnetize or demagnetize small surgical instruments and small steel parts that are over 1.3" in length and 0.125" in diameter. No batteries or electricity required. The box is high quality molded ABS plastic, impact resistant and cadmium free. Dimensions are 2" x 2" x1" The innovative step design lets you control the level of magnetism imposed on your tools.