Learn More
Sklar Instruments Instrument/Drying Tray, Sklar, 13-11/16 in. X 9-13/16 in. X 3/4 in. EA/1.
SDP
Catalog No. 501437146 Shop All Sklar Instruments Products
Encompass_Preferred
Instrument/Drying Tray, Sklar, 13-11/16 in. X 9-13/16 in. X 3/4 in. Instrument/Drying Tray with Rolled Bead Edge, Stainless Steel. Sklar's Instrument/Drying Trays can be used for sterilizing, drying, transporting, and stacking instruments. They are available in a variety of sizes and feature rolled bead edges.
This item is not returnable. View return policy
Catalog No. 50-143-7146 Supplier Sklar Instruments Supplier No. 101883
Instrument/Drying Tray, Sklar, 13-11/16 in. X 9-13/16 in. X 3/4 in. Instrument/Drying Tray with Rolled Bead Edge, Stainless Steel. Sklar's Instrument/Drying Trays can be used for sterilizing, drying, transporting, and stacking instruments. They are available in a variety of sizes and feature rolled bead edges.