Learn More
Med Vet International Instrument ID Tape Roll, Orange, 1/4" x 300", Each
Catalog No. 5000584953
Encompass_Preferred
Instrument ID Roll Tape. These colored tapes are ideal for marking and identifying all kinds of instruments. This allows identification of which instrument belongs to which kit. This will help end the frustration of opening a sterile pack and finding a key instrument missing because it was put in another pack by mistake. The colors are easy to apply and see so the right instrument can be identified every time. Autoclavable. 1/4" x 300" roll.
Catalog No. 50-005-84953 Supplier Med Vet International Supplier No. JOR832F
Instrument ID Roll Tape. These colored tapes are ideal for marking and identifying all kinds of instruments. This allows identification of which instrument belongs to which kit. This will help end the frustration of opening a sterile pack and finding a key instrument missing because it was put in another pack by mistake. The colors are easy to apply and see so the right instrument can be identified every time. Autoclavable. 1/4" x 300" roll.