Learn More
Roboz Surgical Roboz Surgical Instruments Instrument Lubricant, Spray, 8 Oz
SDP
Catalog No. NC1584821
Encompass
This instrument lubricant, also known as instrument milk, is formulated to protect and maintain surgical instruments. The lubricant helps prevent rust, corrosion, and spotting while ensuring smooth operation of hinges and box locks.
  • Prevents rust and corrosion on surgical grade metal
  • Maintains smooth joint and hinge movement
  • Requires no rinsing or wiping after application
  • Convenient spray bottle delivery for easy coverage
  • Shake well before application to dry instruments
Catalog No. NC1584821 Supplier Roboz Surgical Supplier No. IL1000
This instrument lubricant, also known as instrument milk, is formulated to protect and maintain surgical instruments. The lubricant helps prevent rust, corrosion, and spotting while ensuring smooth operation of hinges and box locks.
  • Prevents rust and corrosion on surgical grade metal
  • Maintains smooth joint and hinge movement
  • Requires no rinsing or wiping after application
  • Convenient spray bottle delivery for easy coverage
  • Shake well before application to dry instruments