Learn More
Fine Science Tools Instrument Oil, 50 mL, for Lubricating Instruments
SDP
Catalog No. NC9134687
Encompass
Product: Instrument Oil/ Volume: 50ml/ Features: Physiologically inert oil for lubricating surgical instruments, box locks, ratchets and any overlapping joints/ Unit: Each
Catalog No. NC9134687 Supplier Fine Science Tools Supplier No. 2905500
Product: Instrument Oil/ Volume: 50ml/ Features: Physiologically inert oil for lubricating surgical instruments, box locks, ratchets and any overlapping joints/ Unit: Each