Learn More
Sklar Instruments Vented Tip Guards, Sklar Clear, Disposable, Non-Sterile. It measures 1.5mm x 19mm (1/16 in. x 3/4 in.). PK/100.
SDP
Catalog No. 501184317 Shop All Sklar Instruments Products
Encompass_Preferred
Vented Tip Guards, Sklar, Clear, Disposable, Non-Sterile. Sklar Tip Protectors are used to prevent damage to instrument tips and edges. They are available in a variety of sizes and colors as well as vented or non-vented. This product is clear and vented. It measures 1.5mm x 19mm (1/16 in. x 3/4 in.) and is sold in packs of 100.
Catalog No. 50-118-4317 Supplier Sklar Instruments Supplier No. 961420
Vented Tip Guards, Sklar, Clear, Disposable, Non-Sterile. Sklar Tip Protectors are used to prevent damage to instrument tips and edges. They are available in a variety of sizes and colors as well as vented or non-vented. This product is clear and vented. It measures 1.5mm x 19mm (1/16 in. x 3/4 in.) and is sold in packs of 100.