Learn More
Sklar Instruments Sklar Vented Instrument Protectors, Sklar 25mm, Vented Tip Protectors, Clear, Disposable, Non-Sterile. It measures 25mm x 9.5mm x 1.5mm (1 in. x 3/8 in. x 1/16 in.). PK/100.
SDP
Catalog No. 501184329 Shop All Sklar Instruments Products
Encompass_Preferred
SklarTip Vented Instrument Protectors, Sklar, 25mm, Vented Tip Protectors, Clear, Disposable, Non-Sterile. Sklar Tip Protectors are used to prevent damage to instrument tips and edges. They are available in a variety of sizes and colors as well as vented or non-vented. This product is clear and vented. It measures 25mm x 9.5mm x 1.5mm (1 in. x 3/8 in. x 1/16 in.) and is sold in packs of 100.
Catalog No. 50-118-4329 Supplier Sklar Instruments Supplier No. 961432
SklarTip Vented Instrument Protectors, Sklar, 25mm, Vented Tip Protectors, Clear, Disposable, Non-Sterile. Sklar Tip Protectors are used to prevent damage to instrument tips and edges. They are available in a variety of sizes and colors as well as vented or non-vented. This product is clear and vented. It measures 25mm x 9.5mm x 1.5mm (1 in. x 3/8 in. x 1/16 in.) and is sold in packs of 100.