Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

LumiraDx Instrument for Rapid Detection of SARS-CoV-2

Catalog No. 22181000 Shop All Roche Diagnostics Products
Change view
Click to view available options
Sample Type:
Proteins from SARS to CoV to 2
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Sample Type
22-181-000 Proteins from SARS to CoV to 2
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. 22-181-000 Supplier LumiraDx Supplier No. 10417749160
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Innovative microfluidic technology delivers exceptional clinical performance in minutes, Used with the LumiraDx SARS-CoV-2 Ag Test

  • Features:
    • Innovative, next generation point of care system that combines a small, portable instrument, advanced low-cost test strip and seamless digital connectivity
    • Delivers exceptional point of care clinical performance
    • Simple and easy to use
    • Performance referenced to laboratory test
    • Portable instrument, seamless connectivity
    • One simple, intuitive test workflow
    • Fast results in minutes
    • Nasal swab specimen type
    • Room temperature storage
    • Used with the LumiraDx SARS-CoV-2 Ag Test
  • Notes:
    • Orders require executed contractual agreement
      • Please contact your Fisher Healthcare Account Representative for more information
    • This test has not been FDA cleared or approved; but has been authorized by FDA under EUA for use by authorized laboratories; use by laboratories certified under the CLIA, 42 U.S.C. §263a, that meet requirements to perform moderate, high or waived complexity tests
    • Test authorized for use at the Point of Care (POC), i.e., in patient care settings operating under a CLIA Certificate of Waiver, Certificate of Compliance, or Certificate of Accreditation
    • Authorized only for the detection of proteins from SARS-CoV-2, not for any other viruses or pathogens
    • In USA, test is only authorized for duration of declaration that circumstances exist justifying the authorization of emergency use of in vitro diagnostics for detection and/or diagnosis of the virus that causes COVID-19 under Section 564(b)(1) of the Federal Food, Drug, and Cosmetic Act, 21 U.S.C. § 360bbb-3(b)(1), unless authorization is terminated or revoked sooner
TRUSTED_SUSTAINABILITY

Specifications

Sample Type Proteins from SARS to CoV to 2
Description Used with LumiraDx SARS-CoV-2 Ag Test
Format Portable instrument

Authorized under EUA for use by authorized labs, test has not been FDA cleared or approved

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.