Learn More
Roboz Surgical Roboz Surgical Instrument Roll Pak, Cloth, Large, 1 ea
SDP
Catalog No. NC9908032
Encompass
This large, cloth roll-pack is designed for the organized storage and secure transport of surgical instruments. It helps protect sensitive instruments from damage by providing individual slots for each piece, allowing users to keep instrument types or sets grouped together between uses.
  • Provides eighteen individual slots for organized storage
  • Constructed from durable cloth material
  • Designed for repeated sterilization in an autoclave
  • Measures thirteen by nineteen inches
  • Prevents instrument damage during transport and storage
Catalog No. NC9908032 Supplier Roboz Surgical Supplier No. RS9964
This large, cloth roll-pack is designed for the organized storage and secure transport of surgical instruments. It helps protect sensitive instruments from damage by providing individual slots for each piece, allowing users to keep instrument types or sets grouped together between uses.
  • Provides eighteen individual slots for organized storage
  • Constructed from durable cloth material
  • Designed for repeated sterilization in an autoclave
  • Measures thirteen by nineteen inches
  • Prevents instrument damage during transport and storage