Learn More
Med Vet International Instrument or Spay Pack Tray, J0399A
Catalog No. 5000585346
Encompass_Preferred
Instrument or Spay Pack Tray. Below trays without lids.Top: 101/4" x 63/4" x 2". Bottom: 9" x 51/2" x 2".
Catalog No. 50-005-85346 Supplier Med Vet International Supplier No. JOR399A
Instrument or Spay Pack Tray. Below trays without lids.Top: 101/4" x 63/4" x 2". Bottom: 9" x 51/2" x 2".