Learn More
World Precision Instrument Instrument Sterilization Tray w Silicone Mat, 25.5x15x4cm, Dual Layer
SDP
Catalog No. 502521404
Encompass
Plastic instrument sterilization trays, also known as autoclave trays, are used in healthcare and laboratory settings to hold and transport surgical instruments that require sterilization. Sterilization trays securely hold various instruments, letting you organize and protect them. These trays had snap on lids and silicone mats, making it easy to load instruments into the tray before sterilization and remove them afterward. Plastic sterilization trays are durable and can withstand repeated use, making them a cost-effective choice. The transparent lid allows you to visually inspect you instruments without the need to open the tray. Plastic instrument sterilization trays are compatible with steam autoclaving, ethylene oxide (EO) sterilization, and chemical sterilization. These sterilization trays are available in a variety of sizes.
Catalog No. 50-252-1404 Supplier World Precision Instrument Supplier No. 501728
Plastic instrument sterilization trays, also known as autoclave trays, are used in healthcare and laboratory settings to hold and transport surgical instruments that require sterilization. Sterilization trays securely hold various instruments, letting you organize and protect them. These trays had snap on lids and silicone mats, making it easy to load instruments into the tray before sterilization and remove them afterward. Plastic sterilization trays are durable and can withstand repeated use, making them a cost-effective choice. The transparent lid allows you to visually inspect you instruments without the need to open the tray. Plastic instrument sterilization trays are compatible with steam autoclaving, ethylene oxide (EO) sterilization, and chemical sterilization. These sterilization trays are available in a variety of sizes.