Learn More
Lakeside Manufacturing Inc Instrument Table H-Brace 24x36
Catalog No. 502884442
Encompass_Preferred
Instrument Tables with H-Braces by Lakeside Manufacturing provide superior durability allowing surgeons to hold more supplies and equipment on one shelf. The Lakeside 8391 Instrument Table with H-Brace has (1) top measuring 24" x 36" and a weight capacity of 200-Lb. This Lakeside instrument table's stainless steel top is easy to clean and sanitize. Its 1” (24) square 16-gauge stainless steel legs offer superior strength during transport. This table's all welded construction increases durability and makes surface smoother for cleaning. This table's H-brace adds strength where the cart is most vulnerable to stress. Its 3” (76) locking casters ensure table is stationary when needed. Sits at a comfortable working height of 34" allows easy for access to medical supplies and equipment. 200-Lb. (91kg) weight capacity holds light duty medical supplies and equipment. Reinforcements supply extra security for larger models. Superior lead times. Made in the USA.
Catalog No. 50-288-4442 Supplier Lakeside Manufacturing Inc Supplier No. 8391
Instrument Tables with H-Braces by Lakeside Manufacturing provide superior durability allowing surgeons to hold more supplies and equipment on one shelf. The Lakeside 8391 Instrument Table with H-Brace has (1) top measuring 24" x 36" and a weight capacity of 200-Lb. This Lakeside instrument table's stainless steel top is easy to clean and sanitize. Its 1” (24) square 16-gauge stainless steel legs offer superior strength during transport. This table's all welded construction increases durability and makes surface smoother for cleaning. This table's H-brace adds strength where the cart is most vulnerable to stress. Its 3” (76) locking casters ensure table is stationary when needed. Sits at a comfortable working height of 34" allows easy for access to medical supplies and equipment. 200-Lb. (91kg) weight capacity holds light duty medical supplies and equipment. Reinforcements supply extra security for larger models. Superior lead times. Made in the USA.