Learn More
Lakeside Manufacturing Inc Instrument Table with Shelf, Stainless Steel, 20" x 48", 200-Lb. Capacity
Catalog No. 502884335
Encompass_Preferred
Instrument Tables by Lakeside Manufacturing are a durable choice for surgeons looking for an efficient, organized, and hygienic workspace. The Lakeside 8358 Instrument Table with (1) top and (1) shelf measuring 20" x 48" has a weight capacity of 200-Lb. This Lakeside instrument table's stainless steel top and shelf are easy to clean and sanitize. Its 1” (24) square 16-gauge stainless steel legs offer superior strength during transport. This table's all welded construction increases durability and makes surface smoother for cleaning. Its 3” (76) locking casters ensure table is stationary when needed. Sits at a comfortable working height of 34" allows easy for access to medical supplies and equipment. 200-Lb. (91kg) weight capacity holds light duty medical supplies and equipment. Reinforcements supply extra security for larger models. Endless variations and feature combinations. Superior lead times. Made in the USA.
Catalog No. 50-288-4335 Supplier Lakeside Manufacturing Inc Supplier No. 8358
Instrument Tables by Lakeside Manufacturing are a durable choice for surgeons looking for an efficient, organized, and hygienic workspace. The Lakeside 8358 Instrument Table with (1) top and (1) shelf measuring 20" x 48" has a weight capacity of 200-Lb. This Lakeside instrument table's stainless steel top and shelf are easy to clean and sanitize. Its 1” (24) square 16-gauge stainless steel legs offer superior strength during transport. This table's all welded construction increases durability and makes surface smoother for cleaning. Its 3” (76) locking casters ensure table is stationary when needed. Sits at a comfortable working height of 34" allows easy for access to medical supplies and equipment. 200-Lb. (91kg) weight capacity holds light duty medical supplies and equipment. Reinforcements supply extra security for larger models. Endless variations and feature combinations. Superior lead times. Made in the USA.