Learn More
Fine Science Tools INSTRUMENT TIP PROTECTOR,25/PK
SDP
Catalog No. NC1820771
Encompass
Instrument Tip ProtectorsMaterial:PlastisolAutoclave Safe: YesDimensions: 9.5 x 25.4mmColor: OrangeThese tip protectors are a convenient, color-coded way to protect surgical instrument tips and edges. They provide exceptional protection and are vented to prevent rusting from condensation. The Assorted Pack (Item No. 29060-00) is 5 of each of the 6 available sizes.
Catalog No. NC1820771 Supplier Fine Science Tools Supplier No. 2906006
Instrument Tip ProtectorsMaterial:PlastisolAutoclave Safe: YesDimensions: 9.5 x 25.4mmColor: OrangeThese tip protectors are a convenient, color-coded way to protect surgical instrument tips and edges. They provide exceptional protection and are vented to prevent rusting from condensation. The Assorted Pack (Item No. 29060-00) is 5 of each of the 6 available sizes.